Would you like to log in again?
National D-ELS-1 TriPower Stereomicroscope
Supplier: NATIONAL OPTICAL & SCIENTIFIC SE
National®
D-ELS-1
470222-458EA 313.1 USD InStock
470222-458
National D-ELS-1 TriPower Stereomicroscope
Microscopes
This new, tri-power stereomicroscope is ideal for elementary, middle, and high school grade levels.
  • Magnification: 10x, 20x, and 40x
  • Objectives: 1x, 2x, and 4x
  • Energy-Efficient LED Illumination Lasts up to 50,000 Hours
  • Rechargeable and Cordless
  • Ideal for Elementary, Middle, and High School Grade Levels
This microscope offers three magnifications in one.
One smooth knob adjustment changes the magnification 10x, 20x, or 40x.
Ordering information: This microscope includes vinyl dust cover, rubber eye shields, and a three-year limited warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty