Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Boreal Compound Microscopes with 8" LCD Tablet
Supplier: VWR International
Boreal2
D-EL401 D-EL301
470222-470EA 1049.75 USD InStock
470222-470 470222-468
Boreal Compound Microscopes with 8" LCD Tablet
Microscopes
Use it as a traditional microscope, or view and share live images with the tablet screen.
  • LCD Android Tablet, Eight Inches
  • Screen Resolution: 800×1280
  • Built-in Motic Hub for Wireless Sharing
  • Motic Images Plus Software Included
These microscopes combine high speed imaging technology with our most popular microscopes.
The EM model is coarse focus only and includes 4X, 10X and 40X objectives and gliding stage with stage clips. The HM model has coarse and fine focus, 4X, 10X, 40X and 100X objectives and a built-in mechanical stage. Both models have LED corded illumination.
The Wi-Fi digital tablets transmit live images to iOS or Android devices. Use them as conventional microscopes or share live images with colleagues using Wi-Fi tablets or wireless laptops. The tablet includes preloaded Motic apps.
Ordering information: This kit includes a lifetime manufacturer's warranty on the microscope chassis and a one-year warranty on the tablet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty