Would you like to log in again?
Celestron Microdirect 1080P HDMI Handheld Digital Microscope
Supplier: CELESTRON INTERNATIONAL INC SE
Celestron®
44316
470223-716EA 330.65 USD InStock
470223-716
Celestron Microdirect 1080P HDMI Handheld Digital Microscope
Microscopes
1080p video streaming of microscopic worlds via included HDMI cable directly to your monitor or projector. NO COMPUTER NEEDED!
  • Records high definition still images. Images are saved directly to a Micro SD card (not included)
  • 10x to 220x power magnification
  • 3.5MP high speed sensor has larger pixel sizes that allow more details and a higher refresh rate
We’re taking our handheld digital microscope line to the next level with the MicroDirect 1080p HDMI Handheld Digital Microscope. Perfect for teachers, students, entomologists, forensic scientists, microscopic explorers, and more. This microscope not only offers true 1080p, high definition viewing, but also still image and video recording.
The MicroDirect 1080p HDMI Handheld Digital Microscope streams in amazingly clear 1080p HD video via HDMI cable-directly through a TV monitor or projector-- No computer needed. This microscope can connect to any compatible monitor or projector via HDMI cable for real 1080p HD video streaming at up to 220x magnification. That means high quality viewing of 3D specimens, samples, and even some slides!
Great for the classroom, lab or home, the 3.5MP high speed sensor allows for high definition streaming and still image and video recording. Still images can be saved directly to the onboard Micro SD slot (card not included) while viewing on monitor. The included PC software and USB cable make recording HD videos simple. Use the included fully adjustable stand to steady the microscope to get the perfect image or video.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty