Would you like to log in again?
3B Scientific® SONOtrain™ Ultrasound Vein Phantom
Supplier: 3B Scientific
3B Scientific®
1019652 1019637
470223-770EA 852.3 USD InStock
470223-770 470224-950
3B Scientific® SONOtrain™ Ultrasound Vein Phantom
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
The material of the SONOtrain™ trainer simulates the feel of real soft tissue for carrying out palpations and injections and shows realistic texture and echogenicity on ultrasound images.
  • Easy to use
  • Cost effective
  • Realistic echogenicity
The SONOtrain™ trainer is an excellent teaching tool for basic training and for improving dexterity and hand-eye coordination skills. The ultrasound block has three blood vessels with diameters of 4, 8 and 15 mm with adjustable fluid flow. Punctures and injections are possible, with self-closing injection channels.
Ordering information: An ultrasound device is not part of the delivery content.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty