Would you like to log in again?
Energy Stick
7250
470223-984EA 126.4 USD InStock
470223-984
Energy Stick
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Energize your lessons on electricity.
  • Demonstrate open and closed circuits
  • No danger of shock
  • Colored lights
Safely demonstrate open and closed series circuits without any danger of electric shocks! When the tube is 'turned on,' the red, green, and blue lights light up and it makes a funny buzzing sound. Activating the tube is the fun part - just touch the metal ends with both hands. Your body forms the conducting material that bridges the gap between the two metal strips and closes the circuit!
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty