Would you like to log in again?
Science Take-Out® Lead An Element Of Danger
Supplier: Science Take-Out
STO-148-NOTG STO-148R STO-148 STO-148U
470224-176EA 126.9 USD InStock
470224-176 470224-175 470224-174 470224-178
Science Take-Out® Lead An Element Of Danger
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Follow The Case Of A Young Child With Lead Poisoning
  • Available as a single kit with materials for 1 student or group
  • Unassembled 10 pack for additional savings
  • No additional equipment needed
This complete experiment setup contains all necessary materials and does not require lab equipment. Students will conduct simulated blood tests to determine the child’s blood lead level. They then test samples of dust,
water, soil, and pottery from the child’s home to determine the source of lead. Finally they interpret readings to plan remediation to reduce lead exposure.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty