Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Projection Screen, Eisco Scientific

Supplier: Eisco Scientific
PH0685
470224-378EA 15.59 USD InStock
470224-378
Projection Screen, Eisco Scientific
Educational Materials Physics Educational Materials Optics Learning Activities
Great optical bench accessory.

  • Great accessory for enhancing the functionality of your optical bench
  • 100×75 mm with graph paper screen divided in millimeters
  • Post is about 2.5" tall and has about a ¼" Ø
  • For use with most optical benches

This receiving screen is a valuable component for use with your optical bench. It measures roughly 4×3" (100×75 mm), the graph paper is divided in millimeters. Features a durable white metal frame and rests on an aluminum post.

Delivery information: Millimeter graph paper included.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty