Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco® Accessories Set for Linear Air Track

Supplier: EISCO SCIENTIFIC LLC
Eisco®
PH0362BACC
470224-380EA 159.6 USD InStock
470224-380
Eisco® Accessories Set for Linear Air Track
Educational Materials Physics Educational Materials Mechanics Learning Activities
Accessories for demonstrating properties and laws of motion using linear air tracks.

  • Complete accessory pack for Eisco labs linear air track systems
  • Contains all components required for using the linear air track
  • Use for studying conservation of energy or momentum, elastic and inelastic collisions, Newton's second law, a many other topics
  • Air track not included

The linear air track (not included) provides a nearly frictionless surface with which one can study a multitude of physical processes in ideal conditions.

The air track gliders float on a cushion of air pushed out through small holes in the top of the air track. Using the many included accessories in this kit, experiments can be performed exploring conservation of energy, conservation of momentum, elastic and inelastic collisions, Newton's second law, among many other topics.

Delivery information: This accessory pack contains all of the components necessary for use with Eisco labs linear air track apparatus (not included; 470157-654, 470157-664). It includes 2 sliders, 10 bolt and wing nuts, 4 spring attachments, 4 string attachments, 4 springs, 2 velcro attachment pairs, 2 pulleys with strings, 4 slotted masses with 1 hanger, 8 photogate flags, 8 risers, 6 additional masses, and 2 photogate arms.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty