Would you like to log in again?
3B Scientific® Muscled Hip Joint
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1000177
470224-872EA 581.7 USD InStock
470224-872
3B Scientific® Muscled Hip Joint
Models Human Biology Models
Represented here is the right hip joint of a male with the individual muscles as well as the muscle origins and insertions on the femur and the hip bone.
  • Muscular origins and insertions are raised and color-coded
  • Removable parts: muscle layers can easily be taken off
  • Realistic coloring of bone and muscles
  • 3 year manufacturers warranty
  • Includes 3B Smart Anatomy
For educational purposes, the origin and insertion areas of the muscles have been raised and presented in color (muscle origin = red; muscle insertion = blue) on the hip joint. The hip muscles have been mounted on their corresponding regions of origin and insertion and are removable from the hip joint for a different perspective. Studying the anatomy of the human hip bone is easy with this hip joint and removable muscle. The relationship of the muscular system and skeletal system are represented clearly with this human hip joint model.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty