Would you like to log in again?
3B Scientific® Cancellous Bone
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1009698
470224-878EA 623.9 USD InStock
470224-878
3B Scientific® Cancellous Bone
Models Human Biology Models
The model shows the spongy bone inside the bone.
  • 3 year manufacturers warranty
  • Includes 3B Smart Anatomy
Its filigree architecture is determined by influences such as pressure, bending and torsion. Using innovative micro CT technology, we have managed to reconstruct an exact 3-dimensional copy of a piece of cancellous bone from an original and enlarge it 100 times.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty