Would you like to log in again?
3B Scientific® MICROanatomy™ Tongue
Supplier: AMERICAN 3B SCIENTIFIC SE
MICROanatomy™
1000247
470224-904EA 474.45 USD InStock
470224-904
3B Scientific® MICROanatomy™ Tongue
Models Human Biology Models
Histologic model of the tongue.
  • 3 year manufacturers warranty
  • Includes 3B Smart Anatomy
It comprises a macroscopic view of the tongue in life size (dorsal view) and microscopic views of the various papillae of the tongue (10-20x life size) and of a taste bud (approx. 450x life size). All views are mounted on a base that also features an overview of the sensory and sensitive innervation of the tongue. A unique model for an intensive study of the tongue.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty