Would you like to log in again?
3B Scientific® CT Bronchial Tree
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1000275
470224-910EA 1103.65 USD InStock
470224-910
3B Scientific® CT Bronchial Tree
Models Human Biology Models
This unique CT bronchial tree model with larynx was created on the basis of computer tomography data of a human (male, approx. 40 years).
  • 3 year manufacturers warranty
  • Includes 3B Smart Anatomy
What is special about this procedure is that the natural spatial 3D-relations and the reciprocal location of the segmental bronchi can be preserved and demonstrated in a realistic way. The result is a life-like CT bronchial tree and larynx. This model is a unique way to study the anatomy of the human lungs.
The larynx with hyoid bone and epiglottis and the trachea with primary and lobar bronchi are depicted in one color on the bronchial tree model. The larynx is detachable at the level of the second tracheal cartilage and divisible in the median plane. The epiglottis on the bronchial tree and larynx is mounted flexibly.
The various segmental bronchi are made of elastic material and depicted in various transparent colors so that they are easier to distinguish visually.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty