Would you like to log in again?
3B Scientific® COPD Pathology
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1012788
470224-912EA 211.4 USD InStock
470224-912
3B Scientific® COPD Pathology
Models Human Biology Models
COPD is a chronic lung disease with severe narrowing of the airways.
  • 3 year manufacturers warranty
  • Includes 3B Smart Anatomy
Even administering medication cannot fully cure this narrowing. The model impressively shows the changes to the bronchial tissue (from bottom to top): normal condition, abnormal mucous secretion, thickening of the bronchial mucosa, permanent damage, after administration of medication.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty