Would you like to log in again?
3B Scientific® Breast with Benign Tumor
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1000345
470224-932EA 281.35 USD InStock
470224-932
3B Scientific® Breast with Benign Tumor
Models Human Biology Models
For demonstration of ultrasonic B-image mode.
  • 3 year manufacturers warranty
Model of a woman’s breast made of 3B SKINlike™ silicone with simulated benign tumor for the demonstration of ultrasonic B-image mode with Ultrasonic Echoscope.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty