Would you like to log in again?
3B Scientific® Spinal Injection Trainer
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1017891 1020629 1017892 1020800 1017893
470224-966EA 260.6 USD InStock
470224-966 470224-954 470231-522 470231-516 470231-518
3B Scientific® Spinal Injection Trainer
Models Human Biology Models
This epidural and spinal injection trainer is very impressive thanks to its extremely realistic haptic feedback, and the use of high-quality, hard-wearing materials.
  • Easy to clean
Thanks to close cooperation with epidural anesthesia specialists, incomparably realistic conditions could be recreated. The light, compact structure can be used in a wide variety of ways. The device has low operating costs and is intuitive to use, ensuring lasting, high-level performance.
A summary of the device's main functions:
Epidural anesthesia using the loss-of-resistance and the hanging-drop technique
Spinal anesthesia with realistic resistance of the dura and arachnoid mater with or without a cannula
Fluid-filled spinal canal with realistic outflow rate thanks to the ability to adjust the excess pressure
An epidural catheter can be inserted into the epidural space
Closed water system
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty