Would you like to log in again?
Vibration Generator
Supplier: AMERICAN SCIENTIFIC LLC SE
8420-00
470225-334EA 193.6 USD InStock
470225-334
Vibration Generator
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Mechanical representation of a sine wave signal.
  • The wave drivers allows you to perform experiments with both transverse and longitudinal waves
This model can handle up to 1 Amp and has a unique locking mechanism, which keep the vibrating post secure when it is being stored. The unit can handle signals up to 10 kHz. Couple it with a digital wave generator to produce precise frequencies.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty