Would you like to log in again?
Ward's® Essentials Catenary Arch Kit
Supplier: EISCO SCIENTIFIC LLC
Ward's®
VWRGP00001
470225-366KT 39.99 USD InStock
470225-366
Ward's® Essentials Catenary Arch Kit
Educational Classroom Kits and Activities
Students will explore the Catenary Arch and it's forces.
  • Hands on lab activity
  • Kit aligned to next generation standards MS-PS2-4, MS-PS2-5
  • Instructions and materials included for 1 group
  • Middle school
The Standing Cantenary Arch Kit is composed of seven wooden blocks that assemble to produce the shape of the catenary arch. This kit engages students to learn engineering concepts such as balancing force and load transfer through the assembly of the arch.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty