Would you like to log in again?
Ward's® Essentials Spiral of Theodorus Kit
Supplier: EISCO SCIENTIFIC LLC
Ward's®
VWRGP00006
470225-376KT 13.99 USD InStock
470225-376
Ward's® Essentials Spiral of Theodorus Kit
Educational Classroom Kits and Activities
Learn about triangular relations through hands on kit.
  • Hands on lab activity
  • Kit aligned to next generation standards MS-ETS1-2, MS-ETS1-3
  • Instructions and materials included for 1 group
  • Middle school
Explore triangular relations by studying right triangles and by building the Spiral of Theodorus. These hands-on activities aid in the conceptualization of abstract mathematical ideas. Kit contains 18 wooden triangles, as well as labels, pen and instruction guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty