Would you like to log in again?
EduPrimerTM Alu DNA Profiling Kits
Supplier: GENOSENSOR CORPORATION
3002 3001
470225-430EA 214.55 USD InStock
470225-430 470225-432
EduPrimerTM Alu DNA Profiling Kits
Educational Materials Biology Educational Materials PCR Learning Activities
The EduPrimerTM DNA Profiling kit is designed to introduce students to two dimorphic genes in the Alu family.
  • Determine the distribution of two alleles of the Alu genes within your classroom, and across the world
  • Introduce students to DNA isolation, PCR and DNA Electrophoresis
  • Fast and simple protocol for easy integration into biotechnology lessons
The EduPrimer™ DNA Profiling Kit is designed to introduce students to the principles and applications of PCR, focusing on its real-life use of DNA profiling in forensics and disease research.
Choose between one of two dimorphic genes in the Alu family (TPA-25 or PV92) for a maximum of four shifted DNA bands per student, demonstrating the power of PCR, and teaching the techniques to utilize it.
Ordering information: Kit includes DNA isolation components (lysis buffer/neutralizer), PCR reaction components including master mix (primers and enzymes) and DNA ladder. Kits come with directions to download instructional material and procedures.
Equipment Needed But Not Included: Microcentrifuge, PCR Thermal Cycler, micropipettes and tips, PCR tubes, Microcentrifuge tubes, heat block or water bath, TAE Buffer, DNA Electrophoresis equipment/reagents including transilluminator/light box for analysis.
Delivery information: Kits include perishable materials, please provide a delivery date at check-out.
Packaging: Kits contain reagents only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty