Would you like to log in again?
Fractional Distillation of Crude Oil
Supplier: ALDON CORP SE
Ward's
AD-KK7107-KIT
470225-448KT 49.45 USD InStock
470225-448
Fractional Distillation of Crude Oil
Educational Materials Chemistry Educational Materials Consumer Chemistry Learning Activities
Understand Distillation and Intermolecular Forces with this Real Life Application.
  • Hands on lab activity that illustrates real life process
  • Instructions and materials for 15 lab groups
Crude oil is a complex mixture of many useful hydrocarbons, such as gasoline, kerosene, butane, and motor oil. In order to be of use, these hydrocarbons must first be separated from each other. The process of separation, known as fractional distillation, takes advantage of the fact that these components have varying boiling and condensation points. In this activity, students will complete a fractional distillation experiment to simulate the fractionation of crude oil. As part of this activity students will understand how a difference in boiling points can be utilized to separate a solution with three different components and how intramolecular forces can affect the boiling point of a substance.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty