Would you like to log in again?
Ward's® Chromatography of Amino Acids
Supplier: ALDON CORP SE
Ward's®
AD-KK7108-KIT
470225-450KT 52.65 USD InStock
470225-450
Ward's® Chromatography of Amino Acids
Educational Classroom Kits and Activities
Do you know how to separate proteins?
  • Middle or high school activity
  • Aligned to NGSS Standards PS1.A and PS1.B
  • Contains a teacher's guide and study study guide copymasters
Amino acids are the fundamental building blocks of all proteins. Often times scientists may need to identify the amino acid contained in a protein. One such method for separating and identifying individual amino acids is paper chromatography.
In this activity, students will perform a paper chromatography experiment on three known amino acids, and then use their results to identify the components of an unknown amino acid mixture.
Caution: This product can expose you to chemicals including Methanol/Methyl isobutyl ketone, which is known to the State of California to cause cancer, birth defects and reproductive harm. For more information go to http://www.P65Warnings.ca.gov.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty