Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium Hydroxide

Supplier: ALDON CORP SE
AA0265-3.8L AD-AA0264P-PK/6 AA0273-500ML AA0275-1L AA0264B-2.5LR AA0264-500MLR AA0275-500ML AA0280-500ML AD-AA0264P-2.5L AA0273-30ML AA0274-500ML
470354-384PK 262.4 USD InStock
470354-384 470225-764 470300-208 470300-206 470300-216 470300-204 470300-214 470300-212 470300-210 470225-686 470354-374
Ammonium Hydroxide
Chemicals for Science Education
CAS Number: 1336-21-6
Formula: NH4OH
Density: 0.900 g/mL
Boiling and Freezing Point: 36°C, -77°C
Synonyms: Ammonium Hydroxide Water Solution
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: Corrosive, irritant and toxic.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty