Would you like to log in again?
Linear Diffraction Glasses
Supplier: RAINBOW SYMPHONY INC SE
01616
470225-846PK 18.7 USD InStock
470225-846
Linear Diffraction Glasses
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Make the study of light and color fun and easy
  • Uses a linear diffraction grating
  • Easily see identifiable spectral images
Highly efficient, 500 lines per millimeter, single axis holographic diffraction grating lenses separate light from any source into its spectral components for study and analysis. Recommended for use with Power Supply and Spectrum Gas Tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty