Would you like to log in again?
Ward's Science Essentials® Metamorphic Rocks Collection
Supplier: VWR
WARD470226-466
470226-466EA 26.45 USD InStock
470226-466
Ward's Science Essentials® Metamorphic Rocks Collection
Specimens Rock Specimens Metamorphic Rocks
Introduce your students to rocks formed metamorphism of other rocks.
  • Contains twelve specimens and photo key
  • Perfect for teaching properties of metamorphic rocks
Ward’s Science Essentials
Engage and inspire students to explore in-demand STEM careers with Ward’s Science Essentials. These multi-disciplinary activities kits allow you to easily build STEM concepts into every lesson – with plenty of a-ha moments along the way.
Experience
Create real-world scenarios in the classroom, helping students see the role of STEM in everyday life.
Collaborate
Group-driven activities and challenges foster teamwork and build leadership, communication, and problem-solving skills.
Make Connections
Cross-curricular content reveals tangible examples of how science, technology, engineering and math work together to solve problems.
Delivery information: Collection contains Quartzite, gneiss (2), phyllite, schist (2), slate (2), marble (2), soapstone and serpentinite.
Caution: Due to the nature of this product it may contain asbestos.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty