Would you like to log in again?
Ward's® Essentials The Ins and Outs of Diffusion Kit
Supplier: Avantor
Ward's®
470227-162KT 101.19 USD InStock
470227-162
Ward's® Essentials The Ins and Outs of Diffusion Kit
Educational Classroom Kits and Activities
How big can a cell grow?
  • Hands on lab activity
  • Kit Aligned to Next Generation Science Standard MS-LS1-2
  • Instructions and materials included for 15 groups
  • Middle school
Cell size is determined by how easily a cell can get nutrients and excrete its wastes. Using agar cubes as a model for a cell, students will determine the best cell size to allow waste elimination and nutrient acquisition.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, email or online.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty