Would you like to log in again?
BASIC Stamp Discovery Kit, USB
Supplier: PARALLAX INC.
27807
470227-252KT 181.4 USD InStock
470227-252
BASIC Stamp Discovery Kit, USB
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This module has enough program space, execution speed, and I/O pins for many applications.
  • Send signals with blinking LEDs
  • Sense contact with pushbuttons
  • Read a dial using a potentiometer
  • Measure brightness with a light sensor
The What's a Microcontroller Parts & Text Kit guides you through over 40 hands-on activities designed to provide you with the best possible introduction to the world of BASIC Stamp programming. The text contains clear explanations, defines electronics terms in a friendly manner, and most importantly builds your confidence so you can start building your own electronics inventions. You may use your Board of Education with the other Parts and Text Kits in our education series.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty