Would you like to log in again?
40000 Count Dual Display Handheld LCR Meters
Supplier: B & K PRECISION SE
878B
470227-630EA 398.69 USD InStock
470227-630
40000 Count Dual Display Handheld LCR Meters
Electrical Meters Digital Multimeters
The handheld LCR meters measure inductance, capacitance, and resistance quickly and precisely.
  • 40,000 counts resolution on primary and 10,000 counts resolution on secondary display
  • Fast auto range design for rapid, easy component measurements
  • Data Hold and min/max/average recording
  • USB (Virtual COM) interface
Fast auto ranging and quick measurement configuration such as measurement parameter and test frequency selection make these meters very simple to operate. The meter also includes handy functions such as data hold, Min/Max/Average recording, tolerance sorting, and relative mode.
Measurement data can continuously transfer to a PC via the meter's mini USB interface, using either the provided data logging software or SCPI commands sent from a custom program.
Applications
Passive component troubleshooting
Electronic assembly
Quality control (component sorting)
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty