Would you like to log in again?
Mini Gyroscope
Supplier: EISCO SCIENTIFIC LLC
PH0342
470227-638EA 107.1 USD InStock
470227-638
Mini Gyroscope
Educational Materials Physics Educational Materials Mechanics Learning Activities
Mini version of the older classic Cenco Gyroscope.
  • Carefully balanced
  • All three pairs of pivots are adjustable
  • Complete fitted on stand.
A 7.5cm brass wheel balanced is perfectly balanced with three pairs of adjustable pivots. The central axis has a small hole in it to wind a string around in order to get the gyroscope spinning. Once spinning students can push on the different axis to demonstrate how the applied force creates a torque that is perpendicular to the applied force.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty