Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Specific Gravity/Heat Set

Supplier: EISCO SCIENTIFIC LLC
PH0112ACB
470227-640EA 41.05 USD InStock
470227-640
Specific Gravity/Heat Set
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
A useful set for density, specific heat and gravity.

  • Useful in thermodynamics and density studies
  • Includes instruction manuals
  • Hooks allow easy measurements from a spring scale

Help beginning students conceptually understand density. A cylinder of zinc, copper, aluminum, tin, and steel, each machine stamped with identification, are included in the kit along with a complete instruction manual that gives background on density, specific gravity, and specific heat and contains three inquiry based student activities with capture sheets. Each cylinder has the same 1/2” diameter and mass but a different length.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty