Would you like to log in again?
Magnet Probe
Supplier: EISCO SCIENTIFIC LLC
PH0798MP
470227-642EA 43.35 USD InStock
470227-642
Magnet Probe
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Visualize the shape of the magnetic field around a magnet.
  • Strong plastic construction
Consists of a small 19 mm length alnico bar magnet mounted in freely rotating gimbal. It rotates in 3D so that all x, y and z co-ordinates, three-dimensional magnetic force fields, can be mapped.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty