Would you like to log in again?
SolderMag Magnifier
Supplier: CARSON OPTICAL
SolderMag™
CP-50
470227-878EA 164.65 USD InStock
470227-878
SolderMag Magnifier
Magnifiers
A 1.75X hands free solder station magnifier with LED's.
  • 4.5X spot lens
  • Soldering holder
  • Solder spool dispenser
  • 16 LED's
  • Use a standard AC/DC outlet or 4 AA batteries
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty