Would you like to log in again?
LaMotte® Aquaponics Test Kit
Supplier: LaMotte
Ward's
3637
470228-152EA 349.9 USD InStock
470228-152
LaMotte® Aquaponics Test Kit
Environmental Test Kits
Aquaponics is growing rapidly as a hobby and as an industry as the farm-to-table trend continues to spread.
  • Includes materials to perform 50 tests
  • Detailed instruction manual
Designed for small to midsize systems, this test kit monitors basic water quality for both the aquaculture and hydroponics segments of your system. A detailed, diagrammed instruction manual is provided along with a quick reference lid instruction. Reagents, labware and accessories are mounted in a foam-lined carrying case. Kit is complete with labware, accessories, and reagents to perform approximately 50 repetitions per test factor. Test chemistries and comparators are compatible with fresh, brackish, or salt water systems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty