Would you like to log in again?
Rudiger® Skeleton Hardware Replacement Kits
Supplier: Rüdiger Anatomie
A309.18 A309.16 A309.17 A309.19
470228-160EA 19.1 USD InStock
470228-160 470228-154 470228-158 470228-156
Rudiger® Skeleton Hardware Replacement Kits
Models Human Biology Models
Sets of those hard to find parts.
  • Replace the hardware on older skeletons
  • Matches your existing hardware from Rudiger
Each of these convenient sets allows you to replace a section of hardware for your Rudiger Skeletons. Each kit contains the material to replace all the hardware at one joint.
Arm Kit: Replaces the assembly at one shoulder and contains 1 U-Bracket, 2 Thumbscrews, 1 Threaded Pin, 2 washers and 1 Screw.
Leg Kit: Replaces the assembly at the pelvis to attach 1 leg and contains 1 U-Bracket, 4 Washers, 1 Threaded Pin, 3 Thumbscrews, and 1 nut.
Knee/Elbow Kit: Includes the hardware to reattach at a knee joint and contains 1 eye screw and 1 thumbscrew.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty