Would you like to log in again?
Elephant Toothpaste
Supplier: ALDON CORP SE
IS1007
470228-174KT 17.85 USD InStock
470228-174
Elephant Toothpaste
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
Exciting SAFE Demo for Young Scientists.
  • Aligned to Next Generation science standards (NGSS)
  • Developed for grades 2-5
Check out this fun demonstration that is classroom safe and shows the effects of a catalyst on a chemical reaction. Discussion topics can include basic chemical reactions, phase changes and catalysts.
Ordering information: Includes 150 mL Hydrogen Peroxide 3%, 30 mL Dish Soap, 1 Clear Bottle (8oz), 5 g Yeast, 5 Plastic Cups (5oz), 5 Stir Sticks.
Caution: WARNING CHOKING HAZARD SMALL PART. Not for children under 3 yrs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty