Would you like to log in again?
Density: Lava Lamp
Supplier: ALDON CORP SE
IS1008
470228-176KT 46.9 USD InStock
470228-176
Density: Lava Lamp
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
What is Density?
  • Aligned to Next Generation science standards (NGSS)
  • Developed for grades 2-5
Density is a concept that can be difficult to explain to your elementary and middle school students. Help them visualize and understand density and introduce the idea of polarity as well with this demonstration for the whole classroom.
Ordering information: Includes 5 Plastic Bottles, 1L Vegetable Oil, 25 mL Food Coloring, 10 Alka Seltzer Tablets.
Caution: WARNING CHOKING HAZARD SMALL PART. Not for children under 3 yrs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty