Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SureSeal™ Colored Microcentrifuge Tubes, Sterile
Supplier: MTC BIO, INC.
C2007_WARDS C2000-OB_WARDS C2000-P_WARDS C2000-AST_WARD C2000-Y_WARDS C2002_WARDS C2000-O_WARDS C2007-AST_WARD C2000-G_WARDS C2000-B_WARDS C2000-R_WARDS C2000_WARDS
470228-428PK 45.9 USD InStock
470228-428 470228-430 470228-440 470228-434 470228-444 470228-422 470228-432 470228-442 470228-438 470228-426 470228-436 470228-424
SureSeal™ Colored Microcentrifuge Tubes, Sterile
Tubes Microcentrifuge Tubes
Super-clear polypropylene tubes in superior packaging.
  • Self-standing dispensing bag allows for easy one-handed use
  • 10 Stop-pops™ cap locks provided with all 1.5 and 2.0 ml tubes
  • Manufactured under cleanroom conditions with BioClear™ crystal-clear copolymer
  • RNase, DNase and pyrogen free
  • Smooth rounded lip prevents thumb soreness
SureSeal™ tubes improve sample visualization, safety, stability, and overall ease of use. The oversized tube cap (with frosted marking surface), combined with the unique ring and groove closure enhances the security of the liquid tight seal.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty