Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Resonance Apparatus

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PH0720 PH0720CUP PH0720L PH0720FTUB PH0720TUBE
470229-052EA 161 USD InStock
470229-052 470229-054 470229-056 470229-058 470218-748
Resonance Apparatus
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
With the Resonance Apparatus, students investigate the effect of frequency on wavelength.

  • Durable
  • Inexpensive
  • For Determining the Wavelength and Velocity of Sound
  • Weight: 4.8 lb.
  • Height: 39" after Assembly

An 98cm, transparent, graduated plastic tube is connected to a water reservoir. Both the tube and the water reservoir are securely mounted to a metal support. The water reservoir may be raised or lowered to adjust the air column.

The apparatus weighs 4.8lbs. and is 39" when assembled. The tube is 2.5cm dia. A tuning fork is required.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty