Would you like to log in again?
TeacherGeek Small Motor with Leads and Mount
Supplier: TeacherGeek
1821-02 1821-01
470229-098EA 4.2 USD InStock
470229-098 470229-100
TeacherGeek Small Motor with Leads and Mount
Educational Materials Physics Educational Materials TeacherGeek System Components
Do you have a contraption to power? Here is a simple solution.
  • The motor turns at the perfect speed for most projects
  • Use these for motor powered, cars, boats, scribble bots, and other inventions
This 1.5-3V motor with alligator clip leads that already has a motor mount and adapter pin attached. Also included is a AA single battery holder. Squeeze the alligator clips to attach them to the battery holder terminals. Turn the motor on and off by attaching and detaching the leads. Motor Shaft Diameter: 2.3mm.
Please note: You may need to give the motor shaft a little turn with your fingers, initially, after connecting it to the battery.
Ordering information: Lead colors will vary. Battery not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty