Would you like to log in again?
Glove Dispensers
Supplier: UNITED PRODUCT & INSTRUMENT SE
22250 22150
470229-856EA 48.2 USD InStock
470229-856 470229-858
Glove Dispensers
Glove Dispensers and Glove Box Holders
Always Have Gloves In Reach!
  • Wall mountable, or sits on counter
  • Strong 3/16" clear acrylic
  • Available in 1 or 2 box sizes
Each of these glove box dispensers is made of heavy duty 3/16" thick acrylic for long life. Use the included mounting hardware to attach to a wall to save counter space and you will always know where to find your box of gloves. The dispensers fit all brands of glove boxes, and the clear construction makes it easy to identify sizes printed on the box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty