Would you like to log in again?
Large Inertia Demonstration Model, Eisco Scientific
Supplier: EISCO SCIENTIFIC LLC
LID17
470230-006EA 76.1 USD InStock
470230-006
Large Inertia Demonstration Model, Eisco Scientific
Educational Materials Physics Educational Materials Mechanics Learning Activities
Perfect demonstration of Newton's first law.
  • High quality metal inertia demonstration model
This heavy duty, metal inertia demonstration model is perfect for years of student abuse and classroom use. Its base is mounted on stable rubber feet with adjustable height allowing the platform to be set from 3/4" to 1" high. The 'trigger' then slides the sheet out from under the ball, dropping it into the receptacle upon the top of the post. Can be used for demonstrations in middle school, high school, and introductory college courses.
Delivery information: The unit comes with 0.75" Ø steel ball and attached, dimpled metal plate and set upon a 6" tall post.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty