Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Specific Gravity Cubes

Supplier: EISCO SCIENTIFIC LLC
PH0108E PH0108F PH0108 PH0108A PH0108D
470230-028EA 6.6 USD InStock
470230-028 470230-026 470230-024 470230-022 470230-020
Specific Gravity Cubes
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Specific Gravity Cubes with Hooks.

  • Brass, iron, aluminum and zinc 20mm cubes
  • Hooks

Specific Gravity Cubes with Hooks (Set of 4) This density cube set contains the follow metal cubes: brass, iron, aluminum and zinc. They are .79in (20mm) cubes.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty