Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Density Cubes - lead free

Supplier: EISCO SCIENTIFIC LLC
PH0108PCBLF
470230-034EA 53.2 USD InStock
470230-034
Density Cubes - lead free
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Density Cubes, Set of 4 Metals with Hooks, 1.26" (32 mm) sides, lead free.

  • Set includes: (1) Brass Cube, (1) Aluminum Cube, (1) Copper Cube, (1) Steel Cube
  • Each cube is approximately 3 cm per side, hooks are permanently attached
  • Great for studying density and specific heat without the use of lead
  • Instructions available online
  • Each cube comes in an individual bag and all 4 cubes are placed in a cardboard box

This versatile set is ideal for studying density and specific heat without using lead.  A block of brass, copper, aluminum, and steel are included in the kit along with a complete instruction manual that gives background on density, specific gravity, and specific heat and contains three student activities with capture sheets and a lab practical quiz.  Each mass is 32mm cubed.  The hook on top of the cubes makes it easy to measure mass using a spring scale as well as easy to add and remove masses from boiling water for specific heat experiments.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty