Would you like to log in again?
Metal shapes, Set of 12
Supplier: EISCO SCIENTIFIC LLC
PH0113
470230-046EA 47.9 USD InStock
470230-046
Metal shapes, Set of 12
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
For density determinations, set of twelve in assorted sizes, shapes and masses.
  • 12 objects in assorted shapes, sizes and masses
  • Brass, iron, aluminum, copper, zinc, lead
  • Mass range of 12 - 25 grams
The Labs Set of Metal Blocks is used for density determinations. It includes two of each brass, iron, aluminum, copper, zinc and lead. Masses vary from 12-25 grams.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty