Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Density Set, Polypropylene, Set 12

Supplier: EISCO SCIENTIFIC LLC
PH0113H
470230-048EA 65.75 USD InStock
470230-048
Density Set, Polypropylene, Set 12
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Set of 12 Polypropylene bars of different length but the same diameter.

  • 12 Rods varying in length from 1.5" to 2.625"
  • To determine the density of the material the student plots mass versus volume and the slope therefore is the density (mass per unit volume). Students will need to do all measurements themselves so a balance and ruler are required
  • Base is made from a durable wood coated for years of use
  • Machined from top quality raw materials to produce accurate results and durability of decades of use

A classic experiment for young minds to demonstrate the density does not change regardless of volume and shape. Polypropylene Density Set, 12 Rods, Varying Length with Holder.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty