Would you like to log in again?
LED Array
Supplier: EISCO SCIENTIFIC LLC
PH01LED
470230-066EA 149.4 USD InStock
470230-066
LED Array
Educational Materials Physics Educational Materials Quantum Mechanics Learning Activities
The unit features 11 vertically stacked colors ranging from bright blue to white.
  • Use to study color vs wavelength and/or experimentally determine Planck's Constant h
  • Works well with Labs Direct Vision Spectroscope (product number PH100QA)
  • 11 vertically stacked LED lights
  • Plot wavelength vs threshold voltage to experimentally determine Planck's Constant
  • 4 mm banana plugs/power supply NOT included. Operates using 0-6V continuous DC unit (Labs part # EPR1332)
LED Array for use in topics of diffraction gratings and light, as well as Planck's Constant calculation. The unit features 11 vertically stacked colors ranging from bright blue to white and is a versatile classroom tool.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty