Would you like to log in again?
Friction Block with Hook
Supplier: EISCO SCIENTIFIC LLC
PH0201605
470230-068EA 10.95 USD InStock
470230-068
Friction Block with Hook
Educational Materials Physics Educational Materials Mechanics Learning Activities
Friction blocks are useful for teaching the basics of force and motion.
  • Wooden friction block with c-hook attached at the terminus
  • Use with a spring scale to measure force and mass - all the numbers you need to solve a variety of motion and force problems
  • Hands-on, economic tool for teaching fundamental concepts in physics
  • Students of any age can learn using the friction block
  • Manufactured in the US
This wooden friction block is perfect for introductory studies in force and motion. The block is fitted with a c-hook so that it can be attached to a spring scale. Slide the block along a flat surface, or pull it up an incline. Observe the block by itself, or add weights to the top. When used in a tandem with a spring scale, the friction block is a powerful tool for learning about friction, kinetic and potential energy, acceleration, and force. This block is sourced and manufactured in the United States.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty