Would you like to log in again?
Friction Block Felt with Hook
Supplier: EISCO SCIENTIFIC LLC
PH0201605F
470230-070EA 16.1 USD InStock
470230-070
Friction Block Felt with Hook
Educational Materials Physics Educational Materials Mechanics Learning Activities
Friction blocks are useful for teaching the basics of force and motion.
  • Wooden friction block with c-hook attached at one end
  • Use with a spring scale to measure force and mass - all the numbers you need to solve a variety of motion and force problems
  • Differing pine and felt surfaces allow for comparative study
  • Hands-on, economic tool for teaching fundamental physics concepts to students of any age
  • Manufactured in the US
Friction blocks are useful for teaching the basics of force and motion in a simple, yet effective way. Fitted with a c-hook, the block can be attached to a spring scale to measure incident forces. Slide the block along a flat surface, or pull it up an incline. Observe the block by itself, or add weights to the top. Measure differences in coefficient of friction between the felt and pine surfaces. A friction block used in tandem with a spring scale is a powerful tool for teaching students about friction, kinetic and potential energy, acceleration, and force. This block is sourced and manufactured in the United States.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty