Would you like to log in again?
Plastic Pulley Triple Parallel
Supplier: EISCO SCIENTIFIC LLC
PH0280C
470230-082EA 7.1 USD InStock
470230-082
Plastic Pulley Triple Parallel
Educational Materials Physics Educational Materials Mechanics Learning Activities
Economical pulley for simple machine experiments.
  • Triple plastic pulley
  • Diameter 50 mm
A high quality, triple plastic pulley in parallel, on a steel frame, with hooks at each end carrying three plastic pulleys each of diameter 50 mm. Inner gap of pulley measures 4 mm, overall length is 8.9 cm. Capable of holding over 30 lbs of weight.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty