Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Plastic Pulley Double Tandem 50 and 38 mm

Supplier: EISCO SCIENTIFIC LLC
PH0280D
470230-084EA 8.25 USD InStock
470230-084
Plastic Pulley Double Tandem 50 and 38 mm
Educational Materials Physics Educational Materials Mechanics Learning Activities
Economical pulley for simple machine experiments.

  • Double plastic pulley, in tandem
  • Pulley diameters of 38 and 50 mm

A high quality, double plastic pulley in tandem, on a steel frame, with hooks at each end carrying two plastic pulleys with diameters of 38 and 50 mm respectively. Inner gap of pulley measures 3 mm, overall length is 11.4 cm. Capable of holding over 30 lbs of weight.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty