Would you like to log in again?
Differential Pulley Economy
Supplier: EISCO SCIENTIFIC LLC
PH0283
470230-094EA 9.55 USD InStock
470230-094
Differential Pulley Economy
Educational Materials Physics Educational Materials Mechanics Learning Activities
Economical pulley for simple machine experiments.
  • One piece plastic pulley
  • Economically friendly pricing
  • 20, 40, 60 mm dia. steps
One piece, plastic differential pulley in three steps of 20, 40, 60 mm diameter, mounted with rod.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty